Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-formylphenylboronic acid, 97%, Thermo Scientific™

Catalog No. AC359540050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC359540050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC359540050 Supplier Thermo Scientific Chemicals Supplier No. 359540050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127972-02-5
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD01319044
InChI Key NKKNXLPHCRLBDY-UHFFFAOYSA-N
Synonym 2-methoxy-5-formylphenylboronic acid, 5-formyl-2-methoxyphenyl boronic acid, 5-formyl-2-methoxybenzeneboronic acid, 5-formyl-2-methoxy-phenyl boronic acid, 2-methoxy-5-formylbenzeneboronic acid, 3-borono-4-methoxybenzaldehyde, 3-dihydroxyboranyl-4-methoxybenzaldehyde, boronic acid, 5-formyl-2-methoxyphenyl, pubchem7991, acmc-209bd6
PubChem CID 2734367
IUPAC Name (5-formyl-2-methoxyphenyl)boronic acid
SMILES COC1=CC=C(C=O)C=C1B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 179.97
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2-Methoxy-5-formylphenylboronic acid
Show More Show Less
missing translation for 'healthHazard2' GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.