Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M23115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23115G 5 g
M2311-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23115G Supplier TCI America Supplier No. M23115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127972-00-3
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.98
MDL Number MFCD02179465
InChI Key CSVKZOZMPSRLTC-UHFFFAOYSA-N
PubChem CID 2773485
IUPAC Name (2-methoxy-5-methylphenyl)boronic acid
SMILES COC1=CC=C(C)C=C1B(O)O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Synonym 2-methoxy-5-methylphenyl boronic acid, 2-methoxy-5-methylbenzeneboronic acid, 2-methoxy-5-methyl-phenyl boronic acid, 2-methoxy-5-methylphenyl boranediol, 5-methyl-2-methoxyphenylboronic acid, boronic acid, 2-methoxy-5-methylphenyl, boronic acid, b-2-methoxy-5-methylphenyl, pubchem23333, acmc-1byqj, 2-methoxy-5-methyl-phenylboronic acid
Formula Weight 165.98
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-5-methylphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.