Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-5-nitropyridine 99.0+%, TCI America™
SDP

Catalog No. M130425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M130425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M130425G Supplier TCI America Supplier No. M130425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5446-92-4
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00006263
InChI Key WUPLOZFIOAEYMG-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitro-pyridine, 5-nitro-2-methoxypyridine, 6-methoxy-3-nitropyridine, pyridine, 2-methoxy-5-nitro, methoxy 2--5-nitropyridine, 2-methoxy-5-nitropyridin, pubchem1207, acmc-1art3, 2-methoxy-5-nitro pyridine, 5-nitro-2-methoxy pyridine
PubChem CID 79525
IUPAC Name 2-methoxy-5-nitropyridine
SMILES COC1=CC=C(C=N1)[N+]([O-])=O

Specifications

Melting Point 109°C
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.