Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-6-methylaminopyridine 98.0+%, TCI America™
SDP

Catalog No. M118525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M118525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M118525G Supplier TCI America Supplier No. M118525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88569-83-9
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00191557
InChI Key GXHQLRBOZRJHPZ-UHFFFAOYSA-N
Synonym 2-methoxy-6-methylamino pyridine, 2-methoxy-6-methylaminopyridine, 6-methoxy 2-pyridyl methylamine, 6-methoxy-n-methyl-pyridin-2-amine, pubchem15374, acmc-1bifn, 6-methoxy-2-methylaminopyridine, methoxymethylaminopyridine, 6-methoxy-n-methyl-2-pyridinamine
PubChem CID 7019168
IUPAC Name 6-methoxy-N-methylpyridin-2-amine
SMILES CNC1=CC=CC(OC)=N1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-6-methylaminopyridine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.