Learn More
Enzo Life Sciences 2-Methoxy-antimycin A (1mg)
SDP
Supplier:  Enzo Life Sciences BMLCM1230001
Encompass_Preferred
An analog of antimycin A (mixture of antimycin A1, A2, A3 and A4) which binds to the hydrophobic groove of Bcl-2 and Bcl-XL thereby inducing apoptosis. It displays selective cytotoxicity for Bcl-XL-expressing cells (10µg/ml). Unlike antimycin A it does not exhibit general mitochondrial toxicity. Purity: ≥85%. Solubility: Soluble in DMSO (12 mg/ml) or ethanol (12 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0427