Learn More
Matrix Scientific 2-Methoxy-benzenesulfonyl chloride, 10130-87-7, MFCD01961367, 1g
Supplier: Matrix Scientific 0331821G
Molecular Formula C7H7ClO3S, Molecular Weight 206.65, Melting Point 50-54°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.