Learn More
Cayman Chemical 2-methoxy Estrone, Purity: ≥95%, CAS: 362-08-3, 10 mg
SDP
Catalog No. 502946868
Encompass_Preferred
An active metabolite of estrone; inhibits proliferation of isolated bovine brain-derived capillary endothelial cells (IC50 = 11.5 µM); increases levels of 6-keto PGF1α in HUVECs at 0.001, 0.01, and 0.1 µM; reduces ovariectomy-induced increases in serum cholesterol in rats at 0.5 mg/kg
Catalog No. 50-294-6868 Supplier Cayman Chemical Supplier No. 3415510 MG
An active metabolite of estrone; inhibits proliferation of isolated bovine brain-derived capillary endothelial cells (IC50 = 11.5 µM); increases levels of 6-keto PGF1α in HUVECs at 0.001, 0.01, and 0.1 µM; reduces ovariectomy-induced increases in serum cholesterol in rats at 0.5 mg/kg