Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxy-N-(4-methoxyphenyl)aniline 98.0+%, TCI America™
SDP

Catalog No. M30671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30671G 1 g
M30675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30671G Supplier TCI America Supplier No. M30671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58751-07-8
Molecular Formula C14H15NO2
MDL Number MFCD27956479
Synonym 2,4′-Dimethoxydiphenylamine

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-N-(4-methoxyphenyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.