Learn More
Medchemexpress LLC 2-Methoxybenzaldehyde (o-Anisaldehyde) | 135-02-4 | 99.8% | C8H8O2 | 100 G
SDP
Catalog No. 5000433206
Encompass_Preferred
2-Methoxybenzaldehyde (o-Anisaldehyde) is a bacterial/fungal inhibitor with a BA50 value of 0.19 for Salmonella, suitable for studying bacterial and fungal infectious diseases.
  • Demonstrates anti-Candida effects by inhibiting plasma membrane H+ ATPase, leading to intracellular acidification and Candida cell death.
  • Exhibits an anti-mutagenic effect on 4-nitroquinoline-1-oxide induced mutations in Escherichia coli.
  • Possesses antibacterial activity against Salmonella enterica with a minimum inhibitory concentration (MIC) of 2.5 mg/mL.
Catalog No. 50-004-33206 Supplier Medchemexpress LLC Supplier No. HY77995100G
2-Methoxybenzaldehyde (o-Anisaldehyde) is a bacterial/fungal inhibitor with a BA50 value of 0.19 for Salmonella, suitable for studying bacterial and fungal infectious diseases.
  • Demonstrates anti-Candida effects by inhibiting plasma membrane H+ ATPase, leading to intracellular acidification and Candida cell death.
  • Exhibits an anti-mutagenic effect on 4-nitroquinoline-1-oxide induced mutations in Escherichia coli.
  • Possesses antibacterial activity against Salmonella enterica with a minimum inhibitory concentration (MIC) of 2.5 mg/mL.