Learn More
Medchemexpress LLC 2-Methoxybenzoic acid | 579-75-9 | 152.15 | 100 G
SDP
Catalog No. 5000414749
Encompass_Preferred
2-Methoxybenzoic acid (NSC 3778) is used as an internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves. Another known use is in the synthesis of Benextramine.
  • Human endogenous metabolite
Catalog No. 50-004-14749 Supplier Medchemexpress LLC Supplier No. HYN1393100G
2-Methoxybenzoic acid (NSC 3778) is used as an internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves. Another known use is in the synthesis of Benextramine.
  • Human endogenous metabolite