Learn More
Medchemexpress LLC 2-Methoxybenzoic acid | 579-75-9 | 152.15 | 50 G
SDP
Catalog No. 5000414711
Encompass_Preferred
2-Methoxybenzoic acid is utilized as an internal standard for the simultaneous quantitation of salicylic acid and its putative biosynthetic precursors in cucumber leaves. It is also used in the synthesis of Benextramine. It is for research use only.
  • Internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves
  • Synthesis of Benextramine
  • For research use only
Catalog No. 50-004-14711 Supplier Medchemexpress LLC Supplier No. HYN139350G
Add to Cart
Add to Cart
2-Methoxybenzoic acid is utilized as an internal standard for the simultaneous quantitation of salicylic acid and its putative biosynthetic precursors in cucumber leaves. It is also used in the synthesis of Benextramine. It is for research use only.
  • Internal standard of salicylic acid and its putative biosynthetic precursors in cucumber leaves
  • Synthesis of Benextramine
  • For research use only