Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxycarbazole 98.0+%, TCI America™
SDP

Catalog No. M28175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28175G 5 g
M28171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28175G Supplier TCI America Supplier No. M28175G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6933-49-9
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.237
MDL Number MFCD09033506
InChI Key MDKVPSJRUXOFFV-UHFFFAOYSA-N
PubChem CID 10797986
IUPAC Name 2-methoxy-9H-carbazole
SMILES COC1=CC2=C(C=C1)C3=CC=CC=C3N2

Specifications

Melting Point 239°C
Color White
Quantity 5 g
Formula Weight 197.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxycarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.