Learn More
Apexbio Technology LLC 2-Methoxycinnamaldehyde 1504-74-1 100mg
SDP
Catalog No. 5000093950
Encompass_Preferred
2-Methoxycinnamaldehyde (CAS 1504-74-1) is a natural compound derived from cinnamon investigated for its biomedical properties Studies indicate that it disrupts mitochondrial membrane potential leading to mitochondrial dysfunction This effect is associated with the activation of caspase-3 and caspase-9 resulting in the induction of apoptosis and reduced cellular proliferation Additionally 2-Methoxycinnamaldehyde has been shown to inhibit the migration of human aortic smooth muscle cells in response to platelet-derived growth factor (PDGF) These attributes support its application in research exploring cancer cell apoptosis and vascular smooth muscle cell biology
Catalog No. 50-000-93950 Supplier Apexbio Technology LLC Supplier No. A4627100
2-Methoxycinnamaldehyde (CAS 1504-74-1) is a natural compound derived from cinnamon investigated for its biomedical properties Studies indicate that it disrupts mitochondrial membrane potential leading to mitochondrial dysfunction This effect is associated with the activation of caspase-3 and caspase-9 resulting in the induction of apoptosis and reduced cellular proliferation Additionally 2-Methoxycinnamaldehyde has been shown to inhibit the migration of human aortic smooth muscle cells in response to platelet-derived growth factor (PDGF) These attributes support its application in research exploring cancer cell apoptosis and vascular smooth muscle cell biology