Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methoxycinnamaldehyde | 1504-74-1 | 98.0% | 162.19 g·mol⁻¹ | C10H10O2 | 500 MG
SDP

Catalog No. 5000210141
Encompass_Preferred

2-Methoxycinnamaldehyde is a phenylpropanoid aldehyde supplied as a solid research reagent for in vitro and biochemical studies. It has reported activity against topoisomerase I/II and NF-κB signaling and is provided in small pack sizes for laboratory use.

  • High purity: 98.0% (product data sheet).
  • Molecular formula: C10H10O2; molecular weight: 162.19 g·mol⁻¹.
  • Appearance: solid, white to yellow.
  • Available pack sizes include 500 MG and other small-scale quantities.
  • Storage recommendations: store under nitrogen at -20°C; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-10141 Supplier Medchemexpress LLC Supplier No. HYW046353500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Methoxycinnamaldehyde is a phenylpropanoid aldehyde supplied as a solid research reagent for in vitro and biochemical studies. It has reported activity against topoisomerase I/II and NF-κB signaling and is provided in small pack sizes for laboratory use.

  • High purity: 98.0% (product data sheet).
  • Molecular formula: C10H10O2; molecular weight: 162.19 g·mol⁻¹.
  • Appearance: solid, white to yellow.
  • Available pack sizes include 500 MG and other small-scale quantities.
  • Storage recommendations: store under nitrogen at -20°C; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.