Learn More
Medchemexpress LLC 2-methoxycinnamaldehyde | 1504-74-1 | 99.0% | 162.19 g/mol | C10H10O2 | 5 G
SDP
Catalog No. 5000210688
Encompass_Preferred
2-Methoxycinnamaldehyde is a natural small-molecule aldehyde used for biochemical and cell-based research. It has reported activity including inhibition of topoisomerase I/II and NF-κB signaling, induction of apoptosis, and effects on mitochondrial and lysosomal function. The material is supplied as a white to yellow solid with typical purity near 98.95% and a molecular weight of 162.19 g/mol. Storage recommendations include keeping the solid under nitrogen at -20°C; in solution, store at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Research-grade small molecule for biochemical assays.
  • Reported biological activities include apoptosis induction and signaling inhibition.
  • High chemical purity suitable for analytical and preparative use.
  • Solid physical form facilitates handling and storage.
  • Storage guidance provided for long-term and in-solution stability.
Catalog No. 50-002-10688 Supplier Medchemexpress LLC Supplier No. HYW0463535G
2-Methoxycinnamaldehyde is a natural small-molecule aldehyde used for biochemical and cell-based research. It has reported activity including inhibition of topoisomerase I/II and NF-κB signaling, induction of apoptosis, and effects on mitochondrial and lysosomal function. The material is supplied as a white to yellow solid with typical purity near 98.95% and a molecular weight of 162.19 g/mol. Storage recommendations include keeping the solid under nitrogen at -20°C; in solution, store at -80°C for up to 6 months or at -20°C for up to 1 month.
  • Research-grade small molecule for biochemical assays.
  • Reported biological activities include apoptosis induction and signaling inhibition.
  • High chemical purity suitable for analytical and preparative use.
  • Solid physical form facilitates handling and storage.
  • Storage guidance provided for long-term and in-solution stability.