Learn More
Medchemexpress LLC 2-methoxycinnamaldehyde | 1504-74-1 | MFCD00007001 | 162.19 g/mol | C10H10O2 | 1 G
SDP
Catalog No. 5000210144
Encompass_Preferred
2-Methoxycinnamaldehyde analytical standard for research use. Molecular formula C10H10O2, molecular weight 162.19 g/mol. Store at -20°C under nitrogen; in solvent: -80°C for up to 6 months or -20°C for 1 month (under nitrogen).
  • Analytical standard suitable for analytical and research applications.
  • Supplied in small pack sizes for reference use.
  • Stable when stored under recommended conditions.
  • Intended for research use only; not validated for medical applications.
Catalog No. 50-002-10144 Supplier Medchemexpress LLC Supplier No. HYW0463531G
2-Methoxycinnamaldehyde analytical standard for research use. Molecular formula C10H10O2, molecular weight 162.19 g/mol. Store at -20°C under nitrogen; in solvent: -80°C for up to 6 months or -20°C for 1 month (under nitrogen).
  • Analytical standard suitable for analytical and research applications.
  • Supplied in small pack sizes for reference use.
  • Stable when stored under recommended conditions.
  • Intended for research use only; not validated for medical applications.