Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methoxycinnamaldehyde | 1504-74-1 | MFCD00007001 | 99.0% | 162.19 | C10H10O2 | 2 G
SDP

Catalog No. 5000210729
Encompass_Preferred

2-Methoxycinnamaldehyde is a naturally occurring aromatic aldehyde found in Cinnamomum cassia and used as a research reagent. It has reported biological activities including inhibition of topoisomerase I/II and NF-κB signaling, induction of mitochondrial dysfunction and lysosomal vesiculation, and demonstrated antitumor effects.

  • Cas number: 1504-74-1.
  • Molecular formula: C10H10O2.
  • Molecular weight: 162.19.
  • Purity: 99.0%.
  • Available in small reagent quantities suitable for laboratory research (examples: 100 mg-50 g, including 2 g).

Catalog No. 50-002-10729 Supplier Medchemexpress LLC Supplier No. HYW0463532G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Methoxycinnamaldehyde is a naturally occurring aromatic aldehyde found in Cinnamomum cassia and used as a research reagent. It has reported biological activities including inhibition of topoisomerase I/II and NF-κB signaling, induction of mitochondrial dysfunction and lysosomal vesiculation, and demonstrated antitumor effects.

  • Cas number: 1504-74-1.
  • Molecular formula: C10H10O2.
  • Molecular weight: 162.19.
  • Purity: 99.0%.
  • Available in small reagent quantities suitable for laboratory research (examples: 100 mg-50 g, including 2 g).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.