Learn More
Medchemexpress LLC 2-methoxycinnamaldehyde | 1504-74-1 | MFCD00007001 | 99.0% | 162.19 g/mol | C10H10O2 | 100 MG
SDP
Catalog No. 5000210143
Encompass_Preferred
2-methoxycinnamaldehyde is a naturally occurring cinnamaldehyde derivative isolated from Cinnamomum cassia and used as a research reagent. It is reported to inhibit topoisomerase I and II and NF-κB signaling, to cause mitochondrial dysfunction and lysosomal vesiculation, and to induce DNA damage and apoptosis, with reported antitumor activity in preclinical studies.
  • Isolated from Cinnamomum cassia.
  • Inhibits topoisomerase I and II and NF-κB signaling.
  • Causes mitochondrial dysfunction and induces lysosomal vesiculation.
  • Leads to DNA damage and apoptosis in cellular models.
  • Reported antitumor effects in preclinical studies.
  • Available as an analytical standard with high purity for research use.
Catalog No. 50-002-10143 Supplier Medchemexpress LLC Supplier No. HYW046353100MG
2-methoxycinnamaldehyde is a naturally occurring cinnamaldehyde derivative isolated from Cinnamomum cassia and used as a research reagent. It is reported to inhibit topoisomerase I and II and NF-κB signaling, to cause mitochondrial dysfunction and lysosomal vesiculation, and to induce DNA damage and apoptosis, with reported antitumor activity in preclinical studies.
  • Isolated from Cinnamomum cassia.
  • Inhibits topoisomerase I and II and NF-κB signaling.
  • Causes mitochondrial dysfunction and induces lysosomal vesiculation.
  • Leads to DNA damage and apoptosis in cellular models.
  • Reported antitumor effects in preclinical studies.
  • Available as an analytical standard with high purity for research use.