Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methoxycinnamic acid | 6099-03-2 | 178.18 | 500 G
SDP

Catalog No. 5000414731
Encompass_Preferred

2-Methoxycinnamic acid is a noncompetitive inhibitor of tyrosinase. It is intended for research use only.

  • Purity: 99.94%
  • CAS number: 6099-03-2
  • Molecular weight: 178.18
  • Formula: C10H10O3
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C(O)/C=C/C1=CC=CC=C1OC
  • Structure classification: Phenylpropanoids, Simple Phenylpropanols
  • Initial source: Plants, Ranunculaceae, Pulsatilla chinensis (Bunge) Regel
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-14731 Supplier Medchemexpress LLC Supplier No. HYN1386500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methoxycinnamic acid is a noncompetitive inhibitor of tyrosinase. It is intended for research use only.

  • Purity: 99.94%
  • CAS number: 6099-03-2
  • Molecular weight: 178.18
  • Formula: C10H10O3
  • Appearance: Solid
  • Color: White to off-white
  • SMILES: O=C(O)/C=C/C1=CC=CC=C1OC
  • Structure classification: Phenylpropanoids, Simple Phenylpropanols
  • Initial source: Plants, Ranunculaceae, Pulsatilla chinensis (Bunge) Regel
  • Shipping: Room temperature in continental US
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.