Learn More
Selleck Chemical LLC 2-Methoxyestradiol (2-MeOE2) 10mg 362-07-2 NSC 659853
SDP
Supplier:  Selleck Chemical LLC S123310MG
Encompass_Preferred
2-Methoxyestradiol (2-MeOE2, NSC 659853) depolymerizes microtubules and blocks HIF-1? nuclear accumulation and HIF-transcriptional activity. 2-Methoxyestradiol induces both autophagy and apoptosis in various carcinogenic cell lines. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6131