Learn More
Enzo Life Sciences 2-Methoxyestradiol (10 mg). CAS: 362-07-2
SDP
Catalog No. 502011749
Encompass_Preferred
An endogenous estrogen metabolite with low affinity for the estrogen receptor but which disrupts microtubule function. Potent inhibitor of endothelial cell proliferation and migration. Inhibits bFGF and VEGF-induced corneal neovascularization in mice. Posttranscriptionally downregulates HIF-1α expression. Alternative name: 2-ME. Purity: ≥95% (TLC). Long Term Storage: Ambient.
Catalog No. 50-201-1749 Supplier Enzo Life Sciences Supplier No. BMLS5400010
An endogenous estrogen metabolite with low affinity for the estrogen receptor but which disrupts microtubule function. Potent inhibitor of endothelial cell proliferation and migration. Inhibits bFGF and VEGF-induced corneal neovascularization in mice. Posttranscriptionally downregulates HIF-1α expression. Alternative name: 2-ME. Purity: ≥95% (TLC). Long Term Storage: Ambient.