Learn More
Cayman Chemical 2-Methoxyestradiol, Purity: ≥95%, CAS: 362-07-2, 50 mg
SDP
Catalog No. 502953259
Encompass_Preferred
A natural metabolite formed by CYP450-mediated hydroxylation followed by catechol-O-methyltransferase (COMT) methylation of estradiol; has anticancer agent activity as an angiogenesis inhibitor via the HIF-1α pathway; levels are significantly lower in women with severe pre-eclampsia
Catalog No. 50-295-3259 Supplier Cayman Chemical Supplier No. 1302150 MG
A natural metabolite formed by CYP450-mediated hydroxylation followed by catechol-O-methyltransferase (COMT) methylation of estradiol; has anticancer agent activity as an angiogenesis inhibitor via the HIF-1α pathway; levels are significantly lower in women with severe pre-eclampsia