Learn More
Medchemexpress LLC 2-Methoxyestrone-d4 | 949885-90-9 | 98.0% | 304.42 | 1 MG
SDP
Catalog No. 5000437026
Encompass_Preferred
2-Methoxyestrone-d4 is the deuterium labeled 2-Methoxyestrone. 2-Methoxyestrone is a methoxylated catechol estrogen and metabolite of estrone, with a pKa of 10.81. It can be used as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-37026 Supplier Medchemexpress LLC Supplier No. HY113252S21MG
2-Methoxyestrone-d4 is the deuterium labeled 2-Methoxyestrone. 2-Methoxyestrone is a methoxylated catechol estrogen and metabolite of estrone, with a pKa of 10.81. It can be used as a tracer or an internal standard for quantitative analysis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.