Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017848 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7848 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7848 Supplier MilliporeSigma Supelco Supplier No. PHR15443X1.2ML
Add to Cart
Add to Cart
Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Ignition Point 548°F
CAS 109-86-4
Vapor Pressure 6.17 mmHg (20°C)
Vapor Density 2.62 (vs. air)
Density 0.965 g/mL (at 25°C)
Flash Point 40°C - Closed Cup
Linear Formula CH3 OCH2 CH2 OH
MDL Number MFCD00002867
Melting Point -85°C
Boiling Point 124°C to 125°C
Synonym 2-Methoxyethanol; Ethylene glycol monomethyl ether; Methyl glycol
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 76.09
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.