Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methoxyethoxymethyl Chloride

Catalog No. 3574805G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-748-05G 5 g
35-748-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 35-748-05G Supplier Sigma Aldrich Supplier No. 3574805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

OH-protecting reagent. MEM ethers are stable to a variety of reaction conditions and are selectively cleaved under aprotic conditions in the presence of a wide rangeof OH-protected moieties.

Specifications

CAS 3970-21-6
Density 1.091 g/mL at 25°C
Boiling Point 50°C to 52°C (13 mmHg, lit.)
Molecular Formula C4H9ClO2
Linear Formula CH3OCH2CH2OCH2Cl
Refractive Index n20/D 1.427
MDL Number MFCD00000888
Quantity 5 g
Synonym beta-Methoxyethoxymethyl chloride; MEM-chloride
Molecular Weight (g/mol) 124.57 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.