Learn More
Medchemexpress LLC 2-methoxyethyl 4-((4-(4-chlorophenoxy)-3,5-difluorophenyl)sulfonyl)-3-(hydroxycarbamoyl)piperaz... | 1224964-36-6 | 98.3% | 548.92 | C21H21ClF2N3O8S | 10 MG
SDP
Catalog No. 5000297973
Encompass_Preferred
XL-784 is a small-molecule selective matrix metalloproteinase (MMP) inhibitor provided for research use. It exhibits potent activity across multiple MMP isoforms and is applied in studies of extracellular matrix remodeling, tumor invasion, and metastasis.
  • Selective inhibition of multiple MMPs with reported IC50s in the subnanomolar to micromolar range (e.g., MMP-13 0.56 nM, MMP-2 0.81 nM).
  • Suitable for biochemical and cell-based studies of extracellular matrix and cancer biology.
  • High purity (98.3%) for reliable experimental outcomes.
  • Provided as a solid and as a DMSO solution for flexible dosing.
  • Molecular weight 548.92 g/mol; CAS number 1224964-36-6.
Catalog No. 50-002-97973 Supplier Medchemexpress LLC Supplier No. HY1948510MG
XL-784 is a small-molecule selective matrix metalloproteinase (MMP) inhibitor provided for research use. It exhibits potent activity across multiple MMP isoforms and is applied in studies of extracellular matrix remodeling, tumor invasion, and metastasis.
  • Selective inhibition of multiple MMPs with reported IC50s in the subnanomolar to micromolar range (e.g., MMP-13 0.56 nM, MMP-2 0.81 nM).
  • Suitable for biochemical and cell-based studies of extracellular matrix and cancer biology.
  • High purity (98.3%) for reliable experimental outcomes.
  • Provided as a solid and as a DMSO solution for flexible dosing.
  • Molecular weight 548.92 g/mol; CAS number 1224964-36-6.