Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyethyl acetoacetate, 97%, Thermo Scientific™

Catalog No. AAL1062218
Change view
Click to view available options
Quantity:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1062218 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1062218 Supplier Thermo Scientific Chemicals Supplier No. L1062218

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 22502-03-0
Boiling Point 120°C to 122°C (20mmHg)
Flash Point 103°C (217°F)
Molecular Formula C7H12O4
Quantity 50g
MDL Number MFCD00009648
Beilstein 2079850
Synonym 2-methoxyethyl acetoacetate, 2-methoxyethylacetoacetate, acetoacetic acid 2-methoxyethyl ester, butanoic acid, 3-oxo-, 2-methoxyethyl ester, methoxyethyl acetoacetate, acmc-1cs35, 2-methoxyethyl-3-oxobutanoate, #, 2-methoxy ethyl acetoacetate, 2-methoxyethyl acetoacetate, lonza quality gc
InChI Key PLHCSZRZWOWUBW-UHFFFAOYSA-N
SMILES COCCOC(=O)CC(C)=O
IUPAC Name 2-methoxyethyl 3-oxobutanoate
Molecular Weight (g/mol) 160.17
PubChem CID 89734
Formula Weight 160.17
Percent Purity 97%
Chemical Name or Material 2-Methoxyethyl acetoacetate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.