Learn More
Cayman Chemical 2-Methoxyhydroquinone, Purity: ≥90%, CAS: 824-46-4, 25 g
SDP
Catalog No. 502912059
Encompass_Preferred
A phenolic compound; reduces TNF-α-induced CCL2 production in vitro (IC50 = 64.3 µM); has been used as a precursor in the synthesis of geldanamycin
Catalog No. 50-291-2059 Supplier Cayman Chemical Supplier No. 3653725 G
A phenolic compound; reduces TNF-α-induced CCL2 production in vitro (IC50 = 64.3 µM); has been used as a precursor in the synthesis of geldanamycin