Learn More
Medchemexpress LLC 2-Methoxyhydroquinone | 824-46-4 | 99.6% | 140.14 | 50 G
SDP
Catalog No. 5000454471
Encompass_Preferred
2-Methoxyhydroquinone is a phenolic compound that reduces TNF-α-induced production of chemokine (C-C motif) ligand 2 (CCL2) with an IC50 value of 64.3 μM. It is also a precursor in the synthesis of the Hsp90 inhibitor Geldanamycin.
  • CCL2 Inhibitor
  • Precursor in the synthesis of the Hsp90 inhibitor Geldanamycin
  • Exhibits antiproliferative activity against human A375, A549, HCT-116, HFF-1, MCF-10A, and SK-MEL-2 cells with an EC50 > 100 μM
Catalog No. 50-004-54471 Supplier Medchemexpress LLC Supplier No. HYW01367550G
2-Methoxyhydroquinone is a phenolic compound that reduces TNF-α-induced production of chemokine (C-C motif) ligand 2 (CCL2) with an IC50 value of 64.3 μM. It is also a precursor in the synthesis of the Hsp90 inhibitor Geldanamycin.
  • CCL2 Inhibitor
  • Precursor in the synthesis of the Hsp90 inhibitor Geldanamycin
  • Exhibits antiproliferative activity against human A375, A549, HCT-116, HFF-1, MCF-10A, and SK-MEL-2 cells with an EC50 > 100 μM