Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyisonicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. M20791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20791G 1 g
M20795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20791G Supplier TCI America Supplier No. M20791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105596-63-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD04115718
InChI Key DPNDWFVORIGXQO-UHFFFAOYSA-N
Synonym 2-methoxyisonicotinic acid, 2-methoxy-4-pyridinecarboxylic acid, 2-methoxy-isonicotinic acid, 4-pyridinecarboxylic acid, 2-methoxy, 2-methoxy-isonicotinicacid, 2-methoxy-4-picolinic acid, pubchem9293, ksc179m4j, 4-pyridinecarboxylicacid, 2-methoxy, 2-methyloxy-4-pyridinecarboxylic acid
PubChem CID 819942
IUPAC Name 2-methoxypyridine-4-carboxylic acid
SMILES COC1=NC=CC(=C1)C(=O)O

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 1 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxyisonicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.