Learn More
Medchemexpress LLC 2-Methoxynaphthalene | 93-04-9 | 98.7% | 158.20 | 500 G
SDP
Catalog No. 5000451699
Encompass_Preferred
2-Methoxynaphthalene can be used to investigate the catalytic benefits of delamination, as well as to study alkali metal-mediated manganization (AMMMn) reactions.
Catalog No. 50-004-51699 Supplier Medchemexpress LLC Supplier No. HYW010179500G
2-Methoxynaphthalene can be used to investigate the catalytic benefits of delamination, as well as to study alkali metal-mediated manganization (AMMMn) reactions.