Learn More
Selleck Chemical LLC 2-Methoxy-1,4-naphthoquinone 25mg 2348-82-5
SDP
Supplier:  Selleck Chemical LLC S366125MG
Encompass_Preferred
2-Methoxy-1,4-naphthoquinone, isolated from the leaves of Impatiens glandulifera, specifically suppressed the expression of PKC ?I, ?, and ? in a concentration-dependent manner in Raji cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8415