Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxynicotinic acid, 97%, Thermo Scientific™

Catalog No. KM01841FA
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM01841FA 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM01841FA Supplier Thermo Scientific Chemicals Supplier No. KM01841FA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16498-81-0
Molecular Formula C7H6NO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00085110
InChI Key FTEZJSXSARPZHJ-UHFFFAOYSA-M
Synonym 2-methoxynicotinic acid, 2-methoxy-nicotinic acid, 2-methoxy-3-pyridinecarboxylic acid, 2-methoxy nicotinic acid, 3-pyridinecarboxylic acid, 2-methoxy, t6nj bo1 cvq wwn, maybridge1_004722, 2-methoxynicotinicacid, pubchem20354, acmc-1btxh
PubChem CID 708093
SMILES COC1=NC=CC=C1C([O-])=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 100 g
IUPAC Name 2-methoxypyridine-3-carboxylate
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Methoxynicotinic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.