Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenethyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. M26571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26571G 1 g
M26575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26571G Supplier TCI America Supplier No. M26571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7417-18-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002889
InChI Key XLDRDNQLEMMNNH-UHFFFAOYSA-N
Synonym 2-(2-Hydroxyethyl)anisole, 2-(2-Methoxyphenyl)ethanol
PubChem CID 81909
IUPAC Name 2-(2-methoxyphenyl)ethan-1-ol
SMILES COC1=CC=CC=C1CCO

Specifications

Color Yellow
Boiling Point 134°C
Quantity 1 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxyphenethyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.