Learn More
Medchemexpress LLC 2-Methoxyphenothiazine | 1771-18-2 | 99.6% | 229.30 | 25 G
SDP
Catalog No. 5000366805
Encompass_Preferred
2-Methoxyphenothiazine is a biochemical reagent used in life sciences or organic chemistry research. It is for research use only and not sold to patients.
  • Inhibits erastin-induced ferroptosis in HT-1080 cells with an EC50 of 1.748 μM.
  • Inhibits the 5-lipoxygenase pathway in RBL-1 cells with an IC50 of 0.02 μM.
  • Can enhance the decolorization efficiency of laccase from *Trametes modesta* toward various dyes.
  • Serves as an active component of a catalyst for electrochemical intensified esterification reactions.
Catalog No. 50-003-66805 Supplier Medchemexpress LLC Supplier No. 25GHYW012258
Add to Cart
Add to Cart
2-Methoxyphenothiazine is a biochemical reagent used in life sciences or organic chemistry research. It is for research use only and not sold to patients.
  • Inhibits erastin-induced ferroptosis in HT-1080 cells with an EC50 of 1.748 μM.
  • Inhibits the 5-lipoxygenase pathway in RBL-1 cells with an IC50 of 0.02 μM.
  • Can enhance the decolorization efficiency of laccase from *Trametes modesta* toward various dyes.
  • Serves as an active component of a catalyst for electrochemical intensified esterification reactions.