Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenothiazine 98.0+%, TCI America™
SDP

Catalog No. M082225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M082225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M082225G Supplier TCI America Supplier No. M082225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1771-18-2
Molecular Formula C13H11NOS
Molecular Weight (g/mol) 229.297
MDL Number MFCD00037838
InChI Key DLYKFPHPBCTAKD-UHFFFAOYSA-N
PubChem CID 74490
IUPAC Name 2-methoxy-10H-phenothiazine
SMILES COC1=CC2=C(C=C1)SC3=CC=CC=C3N2

Specifications

Melting Point 188°C
Quantity 25 g
Formula Weight 229.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxyphenothiazine
EINECSNumber (5)-3800
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.