Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Methoxyphenoxy)acetic acid, 98%, Thermo Scientific™

Catalog No. AC442230250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442230250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC442230250 Supplier Thermo Scientific Chemicals Supplier No. 442230250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1878-85-9
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
InChI Key IHONYPFTXGQWAX-UHFFFAOYSA-N
Synonym 2-methoxyphenoxy acetic acid, 2-2-methoxyphenoxy acetic acid, 2-methoxyphenoxyacetic acid, guaiacoxyacetic acid, acetic acid, 2-methoxyphenoxy, o-methoxyphenoxy acetic acid, unii-7xvz1x365a, acide o-methoxyphenoxyacetique french, o-methoxy phenoxyacetic acid, acetic acid, o-methoxyphenoxy
PubChem CID 15882
IUPAC Name 2-(2-methoxyphenoxy)acetic acid
SMILES COC1=CC=CC=C1OCC(=O)O

Specifications

Melting Point 122°C to 123°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material (2-Methoxyphenoxy)acetic acid
Hazard Category GHS Signal Word: Warning
EINECSNumber 217-526-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.