Learn More
Medchemexpress LLC 2-Methoxyphenyl dihydrouracil | 2377643-33-7 | 95.1% | 264.23 g/mol | C12H12N2O5 | 5 G
SDP
Catalog No. 5000280706
Encompass_Preferred
2-Methoxyphenyl dihydrouracil is a small-molecule cereblon (CRBN) ligand used as a synthetic building block in targeted protein degradation research. It is supplied as a purified solid with characterized molecular formula C12H12N2O5 and molecular weight 264.23 g/mol, and is accompanied by analytical documentation and recommended cold, light-protected storage to preserve stability.
  • CRBN ligand useful for PROTAC synthesis
  • High purity for research applications
  • Available in multiple quantities including 5 g
  • Supplied with certificate of analysis
  • Stable when stored refrigerated and protected from light
Catalog No. 50-002-80706 Supplier Medchemexpress LLC Supplier No. HYW2495005G
2-Methoxyphenyl dihydrouracil is a small-molecule cereblon (CRBN) ligand used as a synthetic building block in targeted protein degradation research. It is supplied as a purified solid with characterized molecular formula C12H12N2O5 and molecular weight 264.23 g/mol, and is accompanied by analytical documentation and recommended cold, light-protected storage to preserve stability.
  • CRBN ligand useful for PROTAC synthesis
  • High purity for research applications
  • Available in multiple quantities including 5 g
  • Supplied with certificate of analysis
  • Stable when stored refrigerated and protected from light