Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenylacetonitrile, 98%

Catalog No. p-7027559
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2440018 50 g
AAB2440030 250 g
AAB244000B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2440018 Supplier Thermo Scientific Chemicals Supplier No. B2440018
Only null left
Add to Cart

CAS: 7035-03-2 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7035-03-2
MDL Number MFCD00001902

Specifications

Melting Point 66°C to 70°C
Boiling Point 141°C to 143°C (15 mmHg)
Flash Point 129°C (264°F)
Molecular Formula C9H9NO
Quantity 50 g
UN Number UN3439
Beilstein 2087748
Synonym 2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
InChI Key DWJKILXTMUGXOU-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1CC#N
IUPAC Name 2-(2-methoxyphenyl)acetonitrile
Molecular Weight (g/mol) 147.18
PubChem CID 81496
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 230-314-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.