Learn More
Medchemexpress LLC 2-Methoxyphenylboronic acid | 5720-06-9 | 151.96 | 100 G
SDP
Catalog No. 5000445904
Encompass_Preferred
2-Methoxyphenylboronic acid is a biochemical reagent used in life science research. It functions as a biological material or organic compound.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Appears as a solid
  • Color is white to off-white
  • Stores at -20°C for 3 years or 4°C for 2 years (powder)
  • Stores at -80°C for 6 months or -20°C for 1 month (in solvent)
  • Highly soluble in DMSO (100 mg/mL)
Catalog No. 50-004-45904 Supplier Medchemexpress LLC Supplier No. HYW002569100G
2-Methoxyphenylboronic acid is a biochemical reagent used in life science research. It functions as a biological material or organic compound.
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Appears as a solid
  • Color is white to off-white
  • Stores at -20°C for 3 years or 4°C for 2 years (powder)
  • Stores at -80°C for 6 months or -20°C for 1 month (in solvent)
  • Highly soluble in DMSO (100 mg/mL)