Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methoxyphenylboronic acid | 5720-06-9 | 151.96 | 50 G
SDP

Catalog No. 5000445948
Encompass_Preferred

2-Methoxyphenylboronic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. It is intended for research use only and is not sold to patients.

  • Chemical formula: C7H9BO3
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL); requires sonication
  • Optimal storage for powder at -20°C for 3 years or 4°C for 2 years
  • Optimal storage for in-solvent solutions at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-45948 Supplier Medchemexpress LLC Supplier No. HYW00256950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methoxyphenylboronic acid is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. It is intended for research use only and is not sold to patients.

  • Chemical formula: C7H9BO3
  • Appearance: Solid, white to off-white
  • Soluble in DMSO (100 mg/mL); requires sonication
  • Optimal storage for powder at -20°C for 3 years or 4°C for 2 years
  • Optimal storage for in-solvent solutions at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.