Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methoxypropene

Catalog No. 17464550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-464-550G 50 g
17-464-5250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 17-464-550G Supplier Sigma Aldrich Supplier No. 17464550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to protect adjacent hydroxyls in inositols as bisacetonides, and 1,2-diols in general.

Specifications

CAS 116-11-0
Density 0.753 g/mL at 25°C
Boiling Point 34°C to 36°C
Molecular Formula C4H8O
Refractive Index n20/D 1.382
Linear Formula CH2=C(CH3)OCH3
MDL Number MFCD00014929
Quantity 50 g
Synonym Isopropenyl methyl ether
Molecular Weight (g/mol) 72.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.