Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methoxypyrazine | 3149-28-8 | 100.0% | 110.12 | 25 G
SDP

Catalog No. 5000451502
Encompass_Preferred

2-Methoxypyrazine is an active compound utilized for research in various biochemical studies. This compound is available as a colorless to light yellow liquid and is suitable for both in vitro and in vivo applications, offering reliable performance for scientific investigations.

  • Appearance: liquid
  • Color: colorless to light yellow
  • Molecular formula: C5H6N2O
  • Shipping: room temperature in continental US (may vary elsewhere)
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage for in solvent form: -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: 100 mg/mL in DMSO (908.13 mM), requires sonication

Catalog No. 50-004-51502 Supplier Medchemexpress LLC Supplier No. 25GHYW010562
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methoxypyrazine is an active compound utilized for research in various biochemical studies. This compound is available as a colorless to light yellow liquid and is suitable for both in vitro and in vivo applications, offering reliable performance for scientific investigations.

  • Appearance: liquid
  • Color: colorless to light yellow
  • Molecular formula: C5H6N2O
  • Shipping: room temperature in continental US (may vary elsewhere)
  • Storage for pure form: -20°C for 3 years, 4°C for 2 years
  • Storage for in solvent form: -80°C for 6 months, -20°C for 1 month
  • In vitro solubility: 100 mg/mL in DMSO (908.13 mM), requires sonication

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.