Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,10-phenanthroline 98.0+%, TCI America™
SDP

Catalog No. M25121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M25121G Supplier TCI America Supplier No. M25121G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3002-77-5
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.24
MDL Number MFCD06643855
InChI Key LQZDYPHFVGRHKD-UHFFFAOYSA-N
PubChem CID 4913356
IUPAC Name 2-methyl-1,10-phenanthroline
SMILES CC1=CC=C2C=CC3=CC=CN=C3C2=N1

Specifications

Melting Point 80°C
Color Yellow
Boiling Point 175°C
Quantity 1 g
Formula Weight 194.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1,10-phenanthroline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.