Learn More
Medchemexpress LLC 2-methyl-1,3-cyclohexanedione | 1193-55-1 | MFCD00001587 | 99.9% | 126.15 g·mol⁻¹ | C7H10O2 | 100 G
SDP
Catalog No. 5000300832
Encompass_Preferred
2-Methyl-1,3-cyclohexanedione is an organic diketone used primarily as a synthetic intermediate in pharmaceutical and cosmetic research. It is provided as a laboratory reagent with high chemical purity and a well-defined molecular profile, suitable for condensation, enolate, and other carbonyl chemistry in medicinal chemistry and formulation development.
  • High purity suitable for research applications.
  • Useful as a synthetic intermediate in pharmaceutical synthesis.
  • Applicable in cosmetic ingredient synthesis and formulation studies.
  • Stable diketone structure enables diverse carbonyl chemistry.
  • Available in multiple pack sizes to support scale-up.
Catalog No. 50-003-00832 Supplier Medchemexpress LLC Supplier No. HYW018778100G
2-Methyl-1,3-cyclohexanedione is an organic diketone used primarily as a synthetic intermediate in pharmaceutical and cosmetic research. It is provided as a laboratory reagent with high chemical purity and a well-defined molecular profile, suitable for condensation, enolate, and other carbonyl chemistry in medicinal chemistry and formulation development.
  • High purity suitable for research applications.
  • Useful as a synthetic intermediate in pharmaceutical synthesis.
  • Applicable in cosmetic ingredient synthesis and formulation studies.
  • Stable diketone structure enables diverse carbonyl chemistry.
  • Available in multiple pack sizes to support scale-up.