Learn More
Medchemexpress LLC 2-methyl-1,3-cyclohexanedione | 1193-55-1 | MFCD00001587 | 99.9% | 126.15 g/mol | C7H10O2 | 5 G
SDP
Catalog No. 5000301903
Encompass_Preferred
2-Methyl-1,3-cyclohexanedione is a small organic diketone (C7H10O2; MW 126.15 g/mol) used as a synthetic intermediate in pharmaceutical and fine chemical synthesis. It is supplied as an off-white to light brown solid with high purity suitable for laboratory research and formulation development.
  • Used as a synthetic intermediate in pharmaceutical and organic synthesis.
  • Reported for use in cosmetic ingredient synthesis.
  • High purity (≈99.9%) suitable for research applications.
  • Off-white to light brown solid form, easy to handle.
  • Chemical formula C7H10O2; molecular weight 126.15 g/mol.
  • Stable at room temperature for typical storage; extended stability in solvent under low-temperature storage.
  • Available in multiple research-scale pack sizes for convenience.
Catalog No. 50-003-01903 Supplier Medchemexpress LLC Supplier No. HYW0187785G
2-Methyl-1,3-cyclohexanedione is a small organic diketone (C7H10O2; MW 126.15 g/mol) used as a synthetic intermediate in pharmaceutical and fine chemical synthesis. It is supplied as an off-white to light brown solid with high purity suitable for laboratory research and formulation development.
  • Used as a synthetic intermediate in pharmaceutical and organic synthesis.
  • Reported for use in cosmetic ingredient synthesis.
  • High purity (≈99.9%) suitable for research applications.
  • Off-white to light brown solid form, easy to handle.
  • Chemical formula C7H10O2; molecular weight 126.15 g/mol.
  • Stable at room temperature for typical storage; extended stability in solvent under low-temperature storage.
  • Available in multiple research-scale pack sizes for convenience.