Learn More
Cayman Chemical 2-Methyl-1-butanol, Purity: ≥98%, CAS: 137-32-6, 100 g
SDP
Catalog No. 502942734
Encompass_Preferred
A VOC and an isomer of pentanol; a synthetic intermediate and an organic solvent that has been used in the synthesis of various compounds
Catalog No. 50-294-2734 Supplier Cayman Chemical Supplier No. 40330100 G
A VOC and an isomer of pentanol; a synthetic intermediate and an organic solvent that has been used in the synthesis of various compounds