Learn More
Cayman Chemical 2-Methyl-1-butnol 250g
SDP
Catalog No. 502942735
Encompass_Preferred
A VOC and an isomer of pentanol; a synthetic intermediate and an organic solvent that has been used in the synthesis of various compounds
Catalog No. 50-294-2735 Supplier Cayman Chemical Supplier No. 40330250 G
A VOC and an isomer of pentanol; a synthetic intermediate and an organic solvent that has been used in the synthesis of various compounds