Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1-pentanol 98.5+%, Thermo Scientific™

Catalog No. AC200392500
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC200392500 250 mL
AC200390010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC200392500 Supplier Thermo Scientific Chemicals Supplier No. 200392500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-30-6
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
InChI Key PFNHSEQQEPMLNI-UHFFFAOYSA-N
Synonym 2-methyl-1-pentanol, 1-pentanol, 2-methyl, 2-methylpentanol-1, 2-methylpentanol, sec-amyl carbinol, 2-methylamyl alcohol, +--2-methylpentanol, 2-mpoh, +--2-methyl-1-pentanol, 2-methyl-2-propylethanol
PubChem CID 7745
IUPAC Name 2-methylpentan-1-ol
SMILES CCCC(C)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.82
Infrared Spectrum Authentic
Refractive Index 1.4170 to 1.4190 (20°C, 589nm)
Linear Formula CH3CH2CH2CH(CH3)CH2OH
Quantity 250 mL
Beilstein 01, 409
Formula Weight 102.18
Percent Purity ≥98.50%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-pentanol, 98.5+%
Show More Show Less
EINECSNumber 203-285-1
RTECSNumber SA7175000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.