Learn More
Description
- Glutaric acid monoethyl ester chloride (ethyl glutaryl chloride) is a higher acyl chloride homolog
- Its reaction with calix[4]arene to form ester group-containing calix[4]arenes has been reported.glutaric acid monoethyl ester chloride may be used as a reagent during the multi-step synthesis of lipidic aminoalcohol and diamine derivatives
Specifications
Specifications
| CAS | 73177-67-0 |
| Molecular Formula | C12H18O |
| Linear Formula | C12H18O |
| MDL Number | MFCD00049211 |
| Quantity | 25 mL |
| Molecular Weight (g/mol) | 178.27 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.